Learn More
Kautex™ LDPE Narrow Neck Wash Bottle Closures
Product Code. 15951431
Click to view available options
For Use With (Equipment):
100 mL Bottle
1000 mL Bottle
200 mL Bottle
2000 mL Bottle
250 mL Bottle
500 mL Bottle
Diameter (Metric):
18 mm
25 mm
28 mm
Unit Size:
Each
Pack of 50
Product Code. 15951431
Brand: Kautex™ 2000075992
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Top off Series 301 and Series 312 round, narrow neck bottles with Kautex™ LPDE Narrow Neck Wash Bottle Closures. Their color is natural, which matches the bottle color.
  • LDPE
  • Natural color
  • Round
  • Wash bottle style
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material LDPE
Product Type Wash Bottle Closure
For Use With (Equipment) 200 mL Bottle
Diameter (Metric) 18 mm