Learn More
Vitlab™ LDPE Wash Bottle with Imprint
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Pack of 12
Description
- Narrow/wide-mouth bottles
- Transparent
- Screw cap and spray tube from PP
- Imprinted with Distilled Water in German, English, French and Spanish
- Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Transparent |
| Includes | Screw cap and spray tube |
| Material | LDPE |
| Thread Size (Metric) | GL 45 |
| Closure Included | Yes |
| Height (Metric) | 146 mm |
| Closure Material | Polypropylene |
| Diameter (Metric) | 58 mm |
| Closure Type | Screw Cap |