missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12910454 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 12910454

Brand: Fisher Chemical RP 113 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Quantity 1000 mL
compliance-icons compliance-icons
Product Identifier
  • Lead Acetate Solution. European Pharmacopoeia Ref: 1048103 (RP 113)
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH201-Contains lead. Should not be used on surfaces liable to be chewed or sucked by children.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P314-Get medical advice/attention if you feel unwell.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.