Learn More
Invitrogen™ Lebrikizumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559535
 View more versions of this product
Product Code. 30284188
  • € 500.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lebrikizumab has been used in trials studying the treatment of Asthma, Allergic Asthma, Atopic Dermatitis, Idiopathic Pulmonary Fibrosis, and COPD, Chronic Obstructive Pulmonary Disease.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lebrikizumab Humanized
Recombinant Monoclonal
Unconjugated
MILR1444A; PRO301444
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized