missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LECT2 Antibody (OTI2A11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01632
This item is not returnable.
View return policy
Description
LECT2 Monoclonal specifically detects LECT2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LECT2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
| LECT2 | |
| Human recombinant protein fragment corresponding to amino acids 19-151 of human LECT2(NP_002293) produced in E.coli. | |
| Affinity Purified | |
| RUO | |
| 3950 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI2A11 | |
| Unconjugated | |
| chm2, chm-II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628 | |
| Mouse | |
| 14.5 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction