Learn More
Syngene™ LED Module for Syngene Gbox Chemi System
GREENER_CHOICE
Product Code. 15839962 Shop All Syngene Products
Click to view available options
Wavelength:
465nm
520nm
635nm
740nm
Color:
Blue
Green
Infra Red
Red
Product Type:
LED Module
LED Module, Multiplex
Unit Size:
Each
Product Code. 15839962
Brand: Syngene™ LEDGREENM x 2
Only null left
Add to basket
Add to basket
Syngene™ LED Module for Syngene Gbox Chemi System for western fluorescence
Specifications
Color Green
For Use With (Equipment) Gbox Chemi Systems
Quantity 2
Wavelength 520nm
Product Type LED Module, Multiplex