Learn More
Medline Scientific™ LED Ring Illuminator
Product Code. 12653035
Change view
Click to view available options
For Use With (Equipment):
Steddy Stereo Model Microscopes
Varizoom Stereo Model Microscopes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12653035 Varizoom Stereo Model Microscopes Each
12841583 Steddy Stereo Model Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12653035 Supplier Medline Scientific™ Supplier No. 7062.5001
Used to provide flat uniform lighting of a subject during stereo microscopy.
  • With separate power supply and light intensity control
  • For stereo models only
Specifications
Type Ring Illuminator
Description LED
For Use With (Equipment) Varizoom Stereo Model Microscopes
Includes Separate power source and light intensity control
Wattage 9 w