Learn More
Memmert™ Left Hinged Door for UN260plus
Product Code. 17580006 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN260plus Oven
Unit Size:
Each
Product Code. 17580006
Brand: Memmert™ UN260plusB8
Only null left
Add to basket
Add to basket
Door hinged on the left for use with Memmert Ovens.
  • Hinged on the left
  • Accessory
Specifications
For Use With (Equipment) UN260plus Oven
Type Hinged Door