Learn More
Memmert™ Left Hinged Door for UN55
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN55 Oven
Unit Size:
Each
Description
- Hinged on the left
- Accessory
Specifications
Specifications
| For Use With (Equipment) | UN55 Oven |
| Type | Hinged Door |