missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LEFTY Polyclonal Antibody
GREENER_CHOICE

Product Code. 17109353
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17109353 100 μL 100µL
17109183 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17109353 Supplier Invitrogen™ Supplier No. PA5111432

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AQGDCDPEAP MTEGTRCCRQ EMYIDLQGMK WAENWVLEPP GFLAYECVGT C Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 88%.

This gene encodes a member of the TGF-beta family of proteins. A similar secreted protein in mouse plays a role in left-right asymmetry determination of organ systems during development. Alternative processing of this protein can yield three different products. This gene is closely linked to both a related family member and a related pseudogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEFTY
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LEFTY1
Gene Accession No. O75610
Gene Alias AI450052; Ebaf; endometrial bleeding-associated factor; left right determination factor 1; LEFTA; Leftb; left-right determination factor 1; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor B; lefty; Lefty protein; LEFTY1; lefty-1; LEFTY2; LEFTYA; LEFTYB; Protein lefty-1; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; RGD1561867; Stimulated by retinoic acid gene 3 protein; Stra3; TGFB4; TGF-beta-4; Transforming growth factor beta-4; UNQ278/PRO317
Gene Symbols LEFTY1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human LEFTY. Recombinant protein control fragment (Product #RP-104506).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10637
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.