missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LEFTY Polyclonal Antibody

Description
Immunogen sequence: AQGDCDPEAP MTEGTRCCRQ EMYIDLQGMK WAENWVLEPP GFLAYECVGT C Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 88%.
This gene encodes a member of the TGF-beta family of proteins. A similar secreted protein in mouse plays a role in left-right asymmetry determination of organ systems during development. Alternative processing of this protein can yield three different products. This gene is closely linked to both a related family member and a related pseudogene.
Specifications
Specifications
| Antigen | LEFTY |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | LEFTY1 |
| Gene Accession No. | O75610 |
| Gene Alias | AI450052; Ebaf; endometrial bleeding-associated factor; left right determination factor 1; LEFTA; Leftb; left-right determination factor 1; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor B; lefty; Lefty protein; LEFTY1; lefty-1; LEFTY2; LEFTYA; LEFTYB; Protein lefty-1; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; RGD1561867; Stimulated by retinoic acid gene 3 protein; Stra3; TGFB4; TGF-beta-4; Transforming growth factor beta-4; UNQ278/PRO317 |
| Gene Symbols | LEFTY1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?