missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lefty1/2 Antibody (SR1556), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22230-100ul
This item is not returnable.
View return policy
Description
Lefty1/2 Monoclonal antibody specifically detects Lefty1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Lefty1/2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7044 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| SR1556 | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
| Beta-4 (Endometrial Bleeding-Associated Factor; LEFTY A), EBAF, endometrial bleeding associated factor (left-right determination, Endometrial Bleeding Associated Factor (Left-Right Determination, Factor A;, Endometrial Bleeding-Associated Factor, LEFTA, Left-Right Determination Factor 2, Left-Right Determination Factor A, LEFTY2, LEFTYA, Protein Lefty-2, Protein Lefty-A, TGFB4, TGF-beta-4, Transforming Growth Factor, Transforming Growth Factor Beta-4 | |
| A synthesized peptide derived from human Lefty1/2 (Uniprot #: O00292) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction