missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LEFTY2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30533720
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30533720 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533720 Supplier Invitrogen™ Supplier No. PA5149381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Left-right determination factor 1 and 2 (Lefty1/2) belong to the TGF-beta superfamily of proteins and are responsible for determination of left-right positioning of visceral organs during embryogenesis. Lefty1 acts as a regulator of Lefty2 and NODAL as the lack of the former resulted in bilateral expression of nodal, lefty2 and Pitx2 (a homeobox gene normally expressed on the left side and blocks the signal for leftness). Lefty2 may play a role in endometrial bleeding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEFTY2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene LEFTY2
Gene Accession No. P57785
Gene Alias 6030463A22Rik; AV214969; Ebaf; Ebaf2; endometrial bleeding associated factor; endometrial bleeding-associated factor; LEFTA; Leftb; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor A; LEFTY2; LEFTYA; MGC46222; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; TGFB4; TGF-beta-4; Transforming growth factor beta-4
Gene Symbols LEFTY2
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Lefty-2, Phe78-Leu368.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320202
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.