missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Legionella Pneumophila Lps Antibody (5F4RC), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73187AF594
This item is not returnable.
View return policy
Description
Legionella Pneumophila Lps Monoclonal antibody specifically detects Legionella Pneumophila Lps in Bacteria samples. It is validated for ELISA
Specifications
| Legionella Pneumophila Lps | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
| ELISA | |
| 5F4RC | |
| ELISA | |
| L. pneumophila, Legionellosis | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with membrane fraction of Legionella pneumophila, strain Philadelphia 1. | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction