missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Legionella pneumophila LPS Monoclonal Antibody (5F4)

Description
MA1-21701 detects Legionella pneumophila LPS in Bacteria samples.
The causative agent of what is commonly called Legionnaire's Disease, Legionella pneumophila causes acute pneumonia. The disease can also present itself as an nonpneumonic, influenza-like illness called Pontiac Fever. Legionella is found in aquatic environments where it's growth is supported by amoebae. In human infections Legionella acts as an intracellular parasite and multiplies in alveolar macrophages. The genus Legionella contains 39 recognized species. The immunological diversity of Legionella is due to the existence of a variety of serogroups. Legionella pneumophila has the most with a total of 14 serogroups.
Specifications
Specifications
| Antigen | Legionella pneumophila LPS |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 5F4 |
| Concentration | 5.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | L. pneumophila; Legionellosis |
| Host Species | Mouse |
| Immunogen | Membrane fraction of Legionella pneumophila, strain Philadelphia 1. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?