missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Legumain/Asparaginyl Endopeptidase Antibody (JE35-56), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32528
This item is not returnable.
View return policy
Description
Legumain/Asparaginyl Endopeptidase Monoclonal antibody specifically detects Legumain/Asparaginyl Endopeptidase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Legumain/Asparaginyl Endopeptidase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34 | |
| Recombinant protein within Human Legumain/Asparaginyl Endopeptidase aa 151-350/ 433. (Uniprot: Q99538) | |
| 100 μL | |
| Cancer | |
| 5641 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE35-56 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction