missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LELP1 Antibody (OTI3D5), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72412AF405
This item is not returnable.
View return policy
Description
LELP1 Monoclonal specifically detects LELP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LELP1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LELP1 | |
| Full length recombinant protein of human LELP1 (NP_001010857) produced in E.coli. | |
| 0.1 mL | |
| 149018.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3D5 | |
| Western Blot, Immunohistochemistry | |
| late cornified envelope-like proline-rich 1, novel small proline-rich protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction