missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEMD1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81920JF549
This item is not returnable.
View return policy
Description
LEMD1 Polyclonal antibody specifically detects LEMD1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| LEMD1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| Janelia Fluor 549 | |
| cancer/testis antigen 50, LEM domain containing 1, LEM domain-containing protein 1, LEMP-1 | |
| LEMD1 antibody was raised against a 19 amino acid synthetic peptide near the center of human LEMD1. The immunogen is located within amino acids 90 - 140 of LEMD1. Amino Acid Squence: LDYKPSKGRRWAARAPST | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 93273 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction