missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LEMD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572733
This item is not returnable.
View return policy
Description
Specificity: At least six isoforms of LEMD1 are known to exist; this antibody will detect the longest isoform. LEMD1 antibody is predicted to not cross-react with LEMD2 and LEMD3.
Specifications
| LEMD1 | |
| Polyclonal | |
| Unconjugated | |
| LEMD1 | |
| 4930540I23Rik; 6430514M23Rik; AI428128; Cancer/testis antigen 50; CT50; LEM domain containing 1; LEM domain protein 1; LEM domain-containing protein 1; Lemd1; LEMP-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 213409, 93273 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q14C37, Q68G75 | |
| LEMD1 | |
| LEMD1 antibody for a 19 amino acid synthetic peptide near the center of human LEMD1. The immunogen is within amino acids 90 - 140 of LEMD1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction