missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEMD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 572.00
Specifications
| Antigen | LEMD3 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18476450
|
Novus Biologicals
NBP1-81265 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18405990
|
Novus Biologicals
NBP1-81265-25ul |
25 μL |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LEMD3 Polyclonal antibody specifically detects LEMD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| LEMD3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol | |
| 23592 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| inner nuclear membrane protein Man1, integral inner nuclear membrane protein, LEM domain containing 3, MAN1LEM domain-containing protein 3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QGQAFHLDRRNSPPNSLTPCLKIRNMFDPVMEIGDQWHLAIQEAILEKCSDNDGIVHIAVDKNSREGCVYVKCLSPEYAGKAFKALHGSWFDGKLVTVKYLRLDRYHHRFPQALTSNTPLKPSNKHMNSMSHLRLRT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title