missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEMD3 Antibody (OTI6C10), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71744PECY55
This item is not returnable.
View return policy
Description
LEMD3 Monoclonal specifically detects LEMD3 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications
| LEMD3 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| LEMD3 | |
| Full length recombinant protein of human LEMD3 (NP_055134) produced in HEK293T cells. | |
| 0.1 mL | |
| 23592.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI6C10 | |
| Flow Cytometry | |
| inner nuclear membrane protein Man1, integral inner nuclear membrane protein, LEM domain containing 3, MAN1LEM domain-containing protein 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction