Learn More
Tocris Bioscience™ Lenalidomide 5'-amine
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Lenalidomide 5'-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand with a terminal amine for onward chemistry to generate Protein Degraders with rigid linkers. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 3-(5-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-2,6-piperidinedione |
| CAS | 191732-70-4 |
| Quantity | 50 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C13H13N3O3 |