missing translation for 'onlineSavingsMsg'
Learn More

Lenampicillin hydrochloride, MedChemExpress

Product Code. p-200097511
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473048 10 mg 10mg
16445787 5 mg 5mg
16410026 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16473048 Supplier Medchem Express Supplier No. HY100500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lenampicillin hydrochloride (KBT 1585 hydrochloride) is an orally active prodrug of Ampicillin and is an effective beta-lactam antibacterial agent that inhibits bacterial penicillin-binding proteins (transpeptidase). Lenampicillin hydrochloride has improved absorption and decreased side effects compares to Ampicillin and is applied in the investigation of the suppurative skin and soft tissue infection .

Lenampicillin hydrochloride (KBT 1585 hydrochloride) is an orally active prodrug of Ampicillin and is an effective beta-lactam antibacterial agent that inhibits bacterial penicillin-binding proteins (transpeptidase). Lenampicillin hydrochloride has improved absorption and decreased side effects compares to Ampicillin and is applied in the investigation of the suppurative skin and soft tissue infection .

Specifications

Chemical Name or Material Lenampicillin hydrochloride
Physical Form Solid
CAS 80734-02-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H24ClN3O7S
For Use With (Application) Cancer-programmed cell death
Synonym KBT 1585 hydrochloride
Solubility Information DMSO : 100 mg/mL (200.82 mM; Need ultrasonic)
SMILES O=C([C@@H]1N(C2=O)[C@]([C@@H]2NC([C@@H](C3=CC=CC=C3)N)=O)([H])SC1(C)C)OCC(OC4=O)=C(O4)C.Cl
Molecular Weight (g/mol) 497.95
Formula Weight 497.95
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.