missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LENG1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41357UV
This item is not returnable.
View return policy
Description
LENG1 Polyclonal antibody specifically detects LENG1 in Human samples. It is validated for Western Blot
Specifications
| LENG1 | |
| Polyclonal | |
| Western Blot | |
| leukocyte receptor cluster (LRC) member 1, leukocyte receptor cluster member 1 | |
| The epitope is recognized to a region between residue 214 to 264 of human Leukocyte Receptor Cluster (Lrc) Member 1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79165 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction