missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LENG8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05886-100ul
This item is not returnable.
View return policy
Description
LENG8 Polyclonal antibody specifically detects LENG8 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| LENG8 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| leukocyte receptor cluster (LRC) member 8 | |
| Produced in rabbits immunized with E. coli-derived Human LENG8 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 114823 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction