Learn More
Invitrogen™ LENG8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116606
Description
Positive Samples: U-251MG, Jurkat, Mouse brain, Mouse eye, Rat kidney, Rat brain Immunogen sequence: YYYPMSMYQS YGSPSQYGMA GSYGSATPQQ PSAPQHQGTL NQPPVPGMDE SMSYQAPPQQ LPSAQPPQPS NPPHGAHTLN SGPQPGTAPA TQHSQAGPAT GQAYGPHTYT EPAKPKKGQQ LWNRMKPAPG TGGLKFNIQK RPFAVTTQSF GSNAEGQHSG FGPQPNPEKV QNHSGSSARG NLSGKPDDWP QDMKEYVERC FTACESEEDK DRTEKLLKE.
LENG8 (Leukocyte Receptor Cluster Member 8) is a Protein Coding gene.
Specifications
| LENG8 | |
| Polyclonal | |
| Unconjugated | |
| LENG8 | |
| AW049671; KIAA1932; LENG8; leukocyte receptor cluster (LRC) member 8; leukocyte receptor cluster member 8; Leukocyte receptor cluster member 8 homolog; MGC40108; pp13842 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114823, 232798, 361506 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.98 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8CBY3, Q96PV6 | |
| LENG8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 101-319 of human LENG8 (NP_4431571). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.