missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEPRE1 Antibody (OTI1H3), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72134AF532
This item is not returnable.
View return policy
Description
LEPRE1 Monoclonal specifically detects LEPRE1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| LEPRE1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LEPRE1 | |
| Full length recombinant protein of human LEPRE1 (NP_071751) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1H3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| EC 1.14.11.7, GROS1OI8, Growth suppressor 1, leprecan, Leprecan-1, Leucine- and proline-enriched proteoglycan 1, leucine proline-enriched proteoglycan (leprecan) 1, MGC117314, P3H1LEPRECAN, prolyl 3-hydroxylase 1 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 64175.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction