missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEPRE1 Monoclonal Antibody (OTI1D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505086
This item is not returnable.
View return policy
Description
Basement membrane-associated chondroitin sulfate proteoglycan (CSPG). Has prolyl 3-hydroxylase activity catalyzing the post-translational formation of 3-hydroxyproline in -Xaa-Pro- Gly- sequences in collagens, especially types IV and V. May be involved in the secretory pathway of cells. Has growth suppressive activity in fibroblasts.Specifications
| LEPRE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q32P28 | |
| P3H1 | |
| Full length human recombit protein of human LEPRE1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1D2 | |
| Unconjugated | |
| P3H1 | |
| GROS1, LEPRE1, OI8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64175 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction