missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEPRE1 Monoclonal Antibody (OTI1D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505086S
This item is not returnable.
View return policy
Description
LEPRE1 Monoclonal antibody specifically detects LEPRE1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| LEPRE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BVS4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55781 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1D2 | |
| Unconjugated | |
| RIOK2 | |
| GROS1; Growth suppressor 1; LEPRE1; leprecan; Leprecan-1; leucine- and proline-enriched proteoglycan 1; leucine proline-enriched proteoglycan (leprecan) 1; OI8; P3H1; procollagen-proline 3-dioxygenase; prolyl 3-hydroxylase 1; PSEC0109 | |
| Human recombinant protein fragment corresponding to amino acids 277-552 of human RIOK2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction