Learn More
LEPRE1 Mouse anti-Human, Clone: OTI1D2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505086
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Basement membrane-associated chondroitin sulfate proteoglycan (CSPG). Has prolyl 3-hydroxylase activity catalyzing the post-translational formation of 3-hydroxyproline in -Xaa-Pro- Gly- sequences in collagens, especially types IV and V. May be involved in the secretory pathway of cells. Has growth suppressive activity in fibroblasts.Specifications
| LEPRE1 | |
| Monoclonal | |
| Unconjugated | |
| P3H1 | |
| GROS1, LEPRE1, OI8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64175 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI1D2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q32P28 | |
| P3H1 | |
| Full length human recombit protein of human LEPRE1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.