missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leptin/OB Antibody (11), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05047G
This item is not returnable.
View return policy
Description
Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for ELISA.
Specifications
| Leptin/OB | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LEP | |
| Recombinant human leptin adsorbed onto aluminum hydroxide gel | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Lipid and Metabolism | |
| 3952 | |
| Human | |
| Purified |
| ELISA | |
| 11 | |
| ELISA | |
| FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Clone 11 binds human leptin Cross-reactivity with other species has not been determined | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction