Learn More
Leptin/OB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-611
This item is not returnable.
View return policy
Description
Leptin/OB Polyclonal specifically detects Leptin/OB in Human, Mouse, Rat, Porcine, Avian, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
| Immunohistochemistry, Immunocytochemistry | |
| Human, Mouse, Pig | |
| IgG |
| Polyclonal | |
| Store at -20C. Avoid freeze-thaw cycles. |