Learn More
Leptin/OB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB300-611
Product Code. 18795293
  • € 481.00 / 50µL
Description
Leptin/OB Polyclonal specifically detects Leptin/OB in Human, Mouse, Rat, Porcine, Avian, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry, Immunocytochemistry
Human, Mouse, Pig
IgG
Polyclonal
Store at -20C. Avoid freeze-thaw cycles.