missing translation for 'onlineSavingsMsg'
Learn More

Leptin/OB Mouse anti-Human, Clone: 11, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-05047

 View more versions of this product

Product Code. 18555421

  • € 497.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Leptin/OB
Monoclonal
1 mg/mL
10mM Phosphate (pH 7.4) and 0.5M NaCl
Mouse
Protein A or G purified
RUO
3952
Store at 4°C. Do not freeze.
IgG1 κ
ELISA
11
Unconjugated
FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS
Recombinant human leptin adsorbed onto aluminum hydroxide gel
0.1 mg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.