missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leptin R Antibody (52263) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB867J
This item is not returnable.
View return policy
Description
Leptin R Monoclonal antibody specifically detects Leptin R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Leptin R | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 52263 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
| Mouse myeloma cell line NS0-derived recombinant human Leptin R, Phe22-Asp839 (predicted), Accession # P48357 | |
| 0.1 mL | |
| Signal Transduction | |
| 3953 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction