Learn More
Leptin R Antibody (52263), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB867MFV450
Code produit. 18690600
  • € 684.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Leptin R Monoclonal antibody specifically detects Leptin R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Leptin R
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
0.1 mL
Signal Transduction
3953
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
52263
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295
Mouse myeloma cell line NS0-derived recombinant human Leptin R, Phe22-Asp839 (predicted), Accession # P48357
RUO
Primary
Human
IgG2b