missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leptin R Antibody (55305), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB497PEATT594
This item is not returnable.
View return policy
Description
Leptin R Monoclonal antibody specifically detects Leptin R in Mouse samples. It is validated for Flow Cytometry
Specifications
| Leptin R | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 55305 | |
| Flow Cytometry | |
| CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Leptin R, Leu21-Leu840 (predicted), Accession # P48356 | |
| 0.5 mL | |
| Signal Transduction | |
| 3953 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction