missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leptin R Antibody (mibavademab), PE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28039PE
This item is not returnable.
View return policy
Description
Leptin R Monoclonal antibody specifically detects Leptin R in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Leptin R | |
| Monoclonal | |
| PE | |
| CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
| LEPR/ CD295 | |
| 0.1 mL | |
| Signal Transduction | |
| 3953 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| mibavademab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction