Learn More
Lersivirine, MedChemExpress
Product Code. 16466426
Encompass
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16466426
Brand: Medchem Express HY14267
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Lersivirine (UK-453061) is potent and selective non-nucleoside reverse transcription inhibitor (NNRTI; IC50=119 nM) with excellent efficacy against NNRTI-resistant viruses. Lersivirine exhibits potent antiretroviral activity against wild-type HIV virus and clinically relevant NNRTI-resistant strains.
Lersivirine (UK-453061) is potent and selective non-nucleoside reverse transcription inhibitor (NNRTI
Specifications
Chemical Name or Material Lersivirine
CAS 473921-12-9
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C17H18N4O2
Quantity 100 mg
Synonym UK-453061
Solubility Information DMSO : 50 mg/mL (161.11 mM; Need ultrasonic)
SMILES CCC1=NN(CCO)C(CC)=C1OC2=CC(C#N)=CC(C#N)=C2
Molecular Weight (g/mol) 310.35
Formula Weight 310.35
Percent Purity 98.33%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only