missing translation for 'onlineSavingsMsg'
Learn More

Lesinurad, MedChemExpress

Product Code. 16418997
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16418997

Brand: Medchem Express HY15258

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lesinurad is a URAT1 and OAT inhibitor, is determined to be a substrate for the kidney transporters OAT1 and OAT3 with Km values of 0.85 and 2 μM, respectively.

Lesinurad is a URAT1 and OAT inhibitor, is determined to be a substrate for the kidney transporters OAT1 and OAT3 with Km values of 0.85 and 2 μM, respectively.

Specifications

Chemical Name or Material Lesinurad
Physical Form Solid
CAS 878672-00-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H14BrN3O2S
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym RDEA594
Solubility Information DMSO : 100 mg/mL (247.35 mM; Need ultrasonic)
SMILES BrC1=NN=C(SCC(O)=O)N1C2=C3C(C=CC=C3)=C(C4CC4)C=C2
Molecular Weight (g/mol) 404.28
Formula Weight 404.28
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.