missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LETMD1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06198MFV450
This item is not returnable.
View return policy
Description
LETMD1 Polyclonal antibody specifically detects LETMD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LETMD1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| 1110019O13Rik, cervical cancer 1 protooncogene, Cervical cancer 1 proto-oncogene protein p40, Cervical cancer proto-oncogene 2 protein, DKFZp586A011, HCCR1, HCCR-1, HCCR-2, HCRR-2, LETM1 domain containing 1, LETM1 domain-containing protein 1, regulator of TP53 | |
| Produced in rabbits immunized with E. coli-derived Human LETMD1 fragment. | |
| 0.1 mL | |
| Cancer | |
| 25875 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction