missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LETMD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 220.00 - € 326.00
Specifications
| Antigen | LETMD1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621458
|
Novus Biologicals
NBP3-06198-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638408
|
Novus Biologicals
NBP3-06198-50ul |
50 μg |
€ 220.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LETMD1 Polyclonal antibody specifically detects LETMD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| LETMD1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, pH7.0 | |
| 25875 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 1110019O13Rik, cervical cancer 1 protooncogene, Cervical cancer 1 proto-oncogene protein p40, Cervical cancer proto-oncogene 2 protein, DKFZp586A011, HCCR1, HCCR-1, HCCR-2, HCRR-2, LETM1 domain containing 1, LETM1 domain-containing protein 1, regulator of TP53 | |
| Produced in rabbits immunized with E. coli-derived Human LETMD1 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title