Learn More
Leucomalachite green D5 (phenyl D5), Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Leucomalachite Green phenyl-d5 |
| CAS | 947601-82-3 |
| Molecular Formula | C23 2H5 H21 N2 |
| Quantity | 10 mg |
| Synonym | Benzenamine, 4,4'-(phenyl-2,3,4,5,6-d5-methylene)bis[N,N-dimethyl-, Leucomalachite Green (phenyl-D5) |
| InChI Formula | InChI=1S/C23H26N2/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17,23H,1-4H3/i5D,6D,7D,8D,9D |
| SMILES | [2H]c1c([2H])c([2H])c(C(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C)c([2H])c1[2H] |
| IUPAC Name | 4-[[4-(dimethylamino)phenyl]-(2,3,4,5,6-pentadeuteriophenyl)methyl]-N,N-dimethylaniline |
| Molecular Weight (g/mol) | 335.5 |
| Formula Weight | 335.241 g/mol |