missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leukotriene A4 Hydrolase/LTA4H Antibody (OTI1E9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71189UV
This item is not returnable.
View return policy
Description
Leukotriene A4 Hydrolase/LTA4H Monoclonal specifically detects Leukotriene A4 Hydrolase/LTA4H in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Leukotriene A4 Hydrolase/LTA4H | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LTA4H | |
| Full-length protein expressed in 293T cell transfected with human Leukotriene A4 Hydrolase/LTA4H expression vector. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1E9 | |
| Western Blot, Immunohistochemistry | |
| EC 3.3.2.6, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, leukotriene A-4 hydrolase, LTA4, LTA-4 hydrolase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 4048.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction