missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leukotriene B4 R1 Antibody (203/14F11), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB099Y
This item is not returnable.
View return policy
Description
Leukotriene B4 R1 Monoclonal antibody specifically detects Leukotriene B4 R1 in Human samples. It is validated for Flow Cytometry
Specifications
| Leukotriene B4 R1 | |
| Monoclonal | |
| DyLight 680 | |
| chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 | |
| HeLa cervical epithelial carcinoma cell line transfected with human Leukotriene B4 R1 | |
| 0.5 mL | |
| GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1241 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 203/14F11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction