missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leupaxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23403
This item is not returnable.
View return policy
Description
Leupaxin Polyclonal antibody specifically detects Leupaxin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Leupaxin | |
| Polyclonal | |
| Western Blot 1:1000 | |
| LDLP, LDPL, leupaxin | |
| Leupaxin (N-terminal) synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of human leupaxin. This peptide sequence is highly conserved in rat and mouse leupaxin, and has low homology to other paxillin family proteins. [UniProt# O60711] | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 9404 | |
| Store at -20°C. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction