missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPXN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579610
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Raji whole cell, human PC-3 whole cell, human U87 whole cell, human MDA-MB-453 whole cell, mouse RAW2647 whole cell. Flow: Raji cell.
The product encoded by this gene is preferentially expressed in hematopoietic cells and belongs to the paxillin protein family. Similar to other members of this focal-adhesion-associated adaptor-protein family, it has four leucine-rich LD-motifs in the N-terminus and four LIM domains in the C-terminus. It may function in cell type-specific signaling by associating with PYK2, a member of focal adhesion kinase family. As a substrate for a tyrosine kinase in lymphoid cells, this protein may also function in, and be regulated by, tyrosine kinase activity. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
| LPXN | |
| Polyclonal | |
| Unconjugated | |
| LPXN | |
| 4933402K05Rik; A530083L21Rik; AV278559; LDLP; LDPL; Leupaxin; LPXN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 107321, 9404 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| O60711, Q99N69 | |
| LPXN | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human Leupaxin (115-129aa KKHLPDKQDHKASLD). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction