missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levobunolol hydrochloride, MedChemExpress
Description
Levobunolol (l-Bunolol) hydrochloride is a potent and nonselective β-adrenergic receptor antagonist. Levobunolol hydrochloride is an ocular hypotensive agent and lowers mean intraocular pressure (IOP). Levobunolol hydrochloride can be used for glaucoma and superior oblique myokymia (SOM) research.
Specifications
Specifications
| Chemical Name or Material | Levobunolol hydrochloride |
| CAS | 27912-14-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H26ClNO3 |
| Quantity | 50 mg |
| Synonym | l-Bunolol hydrochloride |
| Solubility Information | DMSO : ≥ 62.5 mg/mL (190.64 mM) ∣H2O : 50 mg/mL (152.51 mM; Need ultrasonic) |
| SMILES | O=C1CCCC2=C1C=CC=C2OC[C@@H](O)CNC(C)(C)C.[H]Cl |
| Molecular Weight (g/mol) | 327.85 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?