Learn More
Levodropropizine, LoGiCal
Product Code. 30462428
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462428 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30462428 Supplier LoGiCal Supplier No. LGCFOR1246.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-(-)-Dropropizine
CAS 99291-25-5
Analyte or Component Names (S)-(-)-Dropropizine
Shipping Condition Room Temperature
Molecular Formula C13H20N2O2
Certifications/Compliance ISO 9001
Synonym Levodropropizine, (2S)-3-(4-Phenylpiperazin-1-yl)propane-1,2-diol
Solution Type Neat
SMILES OC[C@@H](O)CN1CCN(CC1)c2ccccc2
Industry Type Clinical Testing
Molecular Weight (g/mol) 236.31
Quantity 10 mg