Learn More
Levodropropizine, MedChemExpress
Product Code. 16448826
Encompass
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448826 100 mg 100mg
16414637 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16448826 Supplier Medchem Express Supplier No. HYB1895
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MedChemExpress Levodropropizine (DF-526) is a histamine receptor inhibitor, Levodropropizine is an effective and very well tolerated peripheral antitussive drug.
Levodropropizine (DF-526) is a histamine receptor inhibitor, Levodropropizine is an effective and very well tolerated peripheral antitussive drug.
Specifications
Chemical Name or Material Levodropropizine
CAS 99291-25-5
Color White
Purity Grade Notes Research
Molecular Formula C13H20N2O2
Quantity 100 mg
Synonym (S)-(-)-Dropropizine DF-526
Solubility Information DMSO : ≥ 50 mg/mL (211.59 mM) ∣H2O : 10 mg/mL (42.32 mM; Need ultrasonic)
SMILES OC[C@@H](O)CN1CCN(C2=CC=CC=C2)CC1
Molecular Weight (g/mol) 236.31
Formula Weight 236.31
Percent Purity 99.98%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only