Learn More
Levodropropizine, Mikromol™
Product Code. 17963876
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963876 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17963876 Supplier Mikromol Supplier No. MM1246.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Levodropropizine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Levodropropizine
CAS 99291-25-5
Analyte or Component Names Levodropropizine
Shipping Condition Room Temperature
Molecular Formula C13 H20 N2 O2
Certifications/Compliance ISO 17034
Synonym Levodropropizine, (2S)-3-(4-Phenylpiperazin-1-yl)propane-1,2-diol
InChI Formula InChI=1S/C13H20N2O2/c16-11-13(17)10-14-6-8-15(9-7-14)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2/t13-/m0/s1
SMILES OC[C@@H](O)CN1CCN(CC1)c2ccccc2
IUPAC Name (2S)-3-(4-phenylpiperazin-1-yl)propane-1,2-diol
Molecular Weight (g/mol) 236.31
Quantity 250 mg
Formula Weight 236.1525 g/mol